Learn More
Agilent Technologies Inlet septa, bleed and temperature optimized (BTO) 11mm
Catalog No. NC2902128
Encompass
Inlet septa, bleed and temperature optimized (BTO) 11mm
Catalog No. NC2902128 Supplier Agilent Technologies Supplier No. 51834757
Inlet septa, bleed and temperature optimized (BTO) 11mm