Learn More
Trajan Scientific CTC Analytics PAL System Rheodyne Needle Seal, Stainless Steel, 22g, 10 Pack
Catalog No. NC0704367
Encompass
CTC Analytics PAL System Needle Seal, SS, 22g, 10 Pack For Rheodyne Valve, CTC Part Number: PAL NdlSealRRecommEnded For Rheodyne Valves only. For VICI Valco Valves use 100.LCTEF
Catalog No. NC0704367 Supplier Trajan Scientific Supplier No. PAL.NDLSEALR
CTC Analytics PAL System Needle Seal, SS, 22g, 10 Pack For Rheodyne Valve, CTC Part Number: PAL NdlSealRRecommEnded For Rheodyne Valves only. For VICI Valco Valves use 100.LCTEF