Learn More
Heidolph™ Inlet Valve for Laborota 20, Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900932 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-932 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-932 Supplier Heidolph™ Supplier No. 036252020
Add to Cart
Add to Cart
Inlet valve is a component to dispense media manually into evaporating flask.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov