Learn More
Chemglass Life Sciences Inlet Valve, Chem-Cap, High Vacuum, Complete
SDP
Supplier:  Chemglass Life Sciences CG59001
Encompass_Preferred
Inlet Valve, Chem-Cap, High Vacuum, Complete. Designed to be used as a vent or inlet valve for vacuum systems. Can also be used to construct AIRFREE or Schlenk glassware in place of the standard straight bore glass stopcock. This totally grease-free valve does not require a sidearm since the valve has a bore that runs through the plug and an integral machined-on serrated hose connection for attachment to the vacuum or inert gas source. The primary seal is made on the precision tooled seat with the shaft seal being made with a Chem-Cap encased o-ring. The bore is 4mm with an 8mm OD hose connection.
Catalog No. CG59001