Learn More
Medchemexpress LLC Inlexisertib (DCC-3116) | 2543673-19-2 | 99.3% | 535.60 g/mol | C26H36F3N7O2 | 10 MG
SDP
Catalog No. 5000285537
Encompass_Preferred
Inlexisertib is a small-molecule, orally active inhibitor of the ULK1 and ULK2 kinases used in preclinical cancer research to inhibit autophagy and sensitize KRAS-mutant tumors to targeted therapies.
  • Orally active ULK1/2 inhibitor that blocks autophagy.
  • Demonstrated antiproliferative activity in KRAS-mutant cell lines and tumor growth inhibition in non-small cell lung cancer models.
  • Molecular formula C26H36F3N7O2 and molecular weight 535.60 g/mol.
  • High supplied purity (~99.3%).
  • Available in multiple solid pack sizes and as a 10 mM solution in DMSO for in vitro studies.
Catalog No. 50-002-85537 Supplier Medchemexpress LLC Supplier No. HY16069910MG
Inlexisertib is a small-molecule, orally active inhibitor of the ULK1 and ULK2 kinases used in preclinical cancer research to inhibit autophagy and sensitize KRAS-mutant tumors to targeted therapies.
  • Orally active ULK1/2 inhibitor that blocks autophagy.
  • Demonstrated antiproliferative activity in KRAS-mutant cell lines and tumor growth inhibition in non-small cell lung cancer models.
  • Molecular formula C26H36F3N7O2 and molecular weight 535.60 g/mol.
  • High supplied purity (~99.3%).
  • Available in multiple solid pack sizes and as a 10 mM solution in DMSO for in vitro studies.