Learn More
IDEX Health & Science Upchurch INLINE CHECK VALVE ASSEMBLY
Supplier:  IDEX Health & Science Upchurch CV3000
Encompass_Preferred
Idex Pump Check Valve Accessories, Inline Assembly, Diameter Inner: 2.1 mm, For Use With: Pump Check Valve, Includes: Check valve cartridge, flangeless nut and ferrule
Catalog No. NC1920880