Learn More
Euthanex EP-IL60 Inline CO2 Regulator with 0 to 60 PSI Output for use with house gas supply
Catalog No. NC1648971
Encompass
EP-IL60 Inline CO2 Regulator with 0 to 60 PSI Output. For use with house gas supply " not for use with cylinders. Provides consistent set pressure to CO2 equipment when supplied by house source.
Catalog No. NC1648971 Supplier Euthanex Supplier No. EPIL60
EP-IL60 Inline CO2 Regulator with 0 to 60 PSI Output. For use with house gas supply " not for use with cylinders. Provides consistent set pressure to CO2 equipment when supplied by house source.