Learn More
Hach Company Xenosep Inline Column with Sodium Sulfate, 10g, 24/PK
Supplier: Hach Company 2943501
Xenosep Inline Column with Sodium Sulfate used to dewater the hexane extract prior to concentration. 10g. 24 columns per pack.