Learn More
Hach Company Xenosep Inline Column with Sodium Sulfate, 10g, 48/PK
Supplier:  Hach Company 2943500
Encompass
Xenosep Inline Column with Sodium Sulfate, 10g. Used to dewater the hexane extract prior to concentration. 48 columns per pack.
Catalog No. 50-199-6899