Learn More
IDEX Health & Science Upchurch INLINE FILTR HIGH PRESSURE .5U
Supplier:  IDEX Health & Science Upchurch A431
Encompass_Preferred
Idex High-Pressure Solvent Inline Filters, Pore Size: 0.5um, Housing Material: PEEK
Catalog No. NC1920952