Learn More
Cole-Parmer Advantec 357200 Sanitary Stainless Steel In-Line Filter Holder, for 47 mm membranes
Catalog No. NC2195156 Shop All Cole-Parmer Products
Encompass
These sanitary stainless steel holders are for liquid and gas applications. Use for point-of-use filtration on sterile filling machines and for lab or pilot scale sterile filtration applications. The sanitary inlet and outlet connections are non-threaded and easily disassembled for thorough cleaning. Body and support screen are 304 SS with PTFE gasket and FPM/silicone O-rings. Max pressure at inlet is 71 psi (4.9 bar).
This item is not returnable. View return policy
Catalog No. NC2195156 Supplier Cole-Parmer Supplier No. 0664353
Add to Cart
Add to Cart
These sanitary stainless steel holders are for liquid and gas applications. Use for point-of-use filtration on sterile filling machines and for lab or pilot scale sterile filtration applications. The sanitary inlet and outlet connections are non-threaded and easily disassembled for thorough cleaning. Body and support screen are 304 SS with PTFE gasket and FPM/silicone O-rings. Max pressure at inlet is 71 psi (4.9 bar).