Learn More
Ibidi USA In-line Luer Injection Port
SDP
Catalog No. NC0978931
Encompass
An in-line injection port including a silicone septum, used to inject liquieds into a running flow experiment. Adaptable to a tubing of ID 0.5 - 1.6 mm when combined with a Luer Lock Connector Male and a Luer Lock Connector Female. polycarbonate, sterile, 25 pcs./box
This item is not returnable. View return policy
Catalog No. NC0978931 Supplier Ibidi USA Supplier No. 10820
An in-line injection port including a silicone septum, used to inject liquieds into a running flow experiment. Adaptable to a tubing of ID 0.5 - 1.6 mm when combined with a Luer Lock Connector Male and a Luer Lock Connector Female. polycarbonate, sterile, 25 pcs./box