Learn More
Grainger Inline ORP Kit Ryton PPS BNC Flat Tip
Catalog No. 502768153
Encompass_Preferred
The SENSOREX S8200 ORP 10 BNC Inline ORP Sensor is an inline ORP sensor with a 10mm diameter and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and pressure range of 0-100 PSI. The sensor body and process connection are made of PVDF plastic with a platinum band working electrode and Ag/AgCl reference. It has a response time of 95% in less than 30 seconds. The S8200 is designed for continuous monitoring and control of ORP in water treatment systems. It provides stable and accurate ORP measurement.
Catalog No. 50-276-8153 Supplier Grainger Supplier No. 16Z029
The SENSOREX S8200 ORP 10 BNC Inline ORP Sensor is an inline ORP sensor with a 10mm diameter and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and pressure range of 0-100 PSI. The sensor body and process connection are made of PVDF plastic with a platinum band working electrode and Ag/AgCl reference. It has a response time of 95% in less than 30 seconds. The S8200 is designed for continuous monitoring and control of ORP in water treatment systems. It provides stable and accurate ORP measurement.