Learn More
Waters Corp Waters Inline Waste Valve, 1/4-28 Thread, for ACQUITY FTN Sample Managers
Catalog No. 50791794 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement inline waste valve designed with a 1/4-28 thread, specifically for use with ACQUITY FTN Sample Managers.
  • Utilized with ACQUITY FTN Sample Managers.
  • 1/4-28 thread.
  • System type is UPLC.
  • Replacement schedule is as required.
  • One unit per package.
Catalog No. 50-791-794 Supplier Waters Corp Supplier No. 410003180
This is a replacement inline waste valve designed with a 1/4-28 thread, specifically for use with ACQUITY FTN Sample Managers.
  • Utilized with ACQUITY FTN Sample Managers.
  • 1/4-28 thread.
  • System type is UPLC.
  • Replacement schedule is as required.
  • One unit per package.