Learn More
Aviva Systems Biology Corp INMT Antibody - N-terminal region (ARP48714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INMT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human INMT. Alternate names: MGC125940, MGC125941ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-59894