Learn More
Binder Inc Inner door divided CB 170
SDP
Supplier:  Binder Inc 86120030
Encompass_Preferred
Inner door, divided - Cell therapy compartmentalization, consisting of 6 inner doors and three shelf levels with one partition wall each - optional for BINDER CO2 incubator
Catalog No. 86-120-030