Learn More
BINDER™ Inner Door, Divided
SDP
Catalog No. 5000217502 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
CB 170/CBF 170 Left-hinged Door
CB 170/CBF 170 Right-hinged Door
CB 260/CBF 260
CB 56
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-002-17502 CB 170/CBF 170 Right-hinged Door
50-002-17501 CB 56
50-002-17509 CB 170/CBF 170 Left-hinged Door
50-002-17520 CB 260/CBF 260
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-002-17502 Supplier BINDER™ Supplier No. 80122024
Only null left
Add to Cart
Add to Cart
Cell therapy compartmentalization, consisting of 4, 6, 8 inner doors, 6 inner door for a left-hand door and two shelf levels with one partition wall each
A divided inner door lets you open only a small section of the incubator instead of the whole chamber, which greatly protects the internal atmosphere and the cells. This design keeps temperature, CO2, humidity, and (if used) O2 much more stable during routine access, which improves cell health and experimental reproducibility.
  • Opening just one segment limits how much warm, humid, CO2‐rich air escapes, so the incubator recovers to setpoints faster after each door opening.
  • More stable conditions from fewer fluctuations in temperature and gases support more consistent growth of sensitive cells, especially in shared incubators that are opened many times per day.
  • Because each opening affects a smaller volume, the incubator needs less CO2 (and N2 in hypoxic mode) to return to the desired atmosphere, which can cut gas use by 40 to 80% compared with a single large inner door.
  • Lower gas consumption extends cylinder lifetime, reduces cost and workload for changing bottles, and helps meet sustainability goals.
  • Segmented doors also separate different shelves or compartments, helping to spatially isolate different users or experiments (for example, clinical or cell‐therapy work) within the same incubator.
Specifications
For Use With (Equipment) CB 170/CBF 170 Right-hinged Door
No. of Places 6
No. of Shelves 2
Type Inner Door