Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Electrolyte for Carbon Dioxide Gas Sensitive Electrode

Catalog No. 01913232 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-913-232 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-913-232 Supplier Mettler Toledo™ Supplier No. 51340038
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Carbon Dioxide Gas Sensitive Electrode.

  • Quantity: 50mL
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description Electrolyte for carbon dioxide CO2, 25mL
For Use With (Equipment) Carbon Dioxide Gas Sensitive Electrode
Type Electrolyte
Filling Solution Type Electrolyte
Packaging 25 mL
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.