Learn More
MSC Baldwin Filters Radial Seal Inner Air Element, 174.6 mm OD, 276.2 mm Height, 1 ea
Catalog No. 5000509890
Encompass
This radial seal inner air filter element is designed for heavy-duty applications, specifically compatible with John Deere tractors. It serves as an internal secondary filter to ensure optimal engine protection.
  • Heavy-duty radial seal gasket and end cap resist degradation under extreme temperatures, vibration, and restriction changes
  • Flexible outer edges allow for easy installation and removal in housings with minimal clearance
  • PermaPleat construction ensures even pleat spacing to prevent bunching and extend service life
  • Spiral glue binding provides increased pleat stability for maximum filtration media effectiveness
  • Designed for one-end installation with a 134.9 mm (5 5/16 in.) inside diameter
Catalog No. 50-005-09890 Supplier MSC Supplier No. 55874101
This radial seal inner air filter element is designed for heavy-duty applications, specifically compatible with John Deere tractors. It serves as an internal secondary filter to ensure optimal engine protection.
  • Heavy-duty radial seal gasket and end cap resist degradation under extreme temperatures, vibration, and restriction changes
  • Flexible outer edges allow for easy installation and removal in housings with minimal clearance
  • PermaPleat construction ensures even pleat spacing to prevent bunching and extend service life
  • Spiral glue binding provides increased pleat stability for maximum filtration media effectiveness
  • Designed for one-end installation with a 134.9 mm (5 5/16 in.) inside diameter