Learn More
MSC Baldwin Radial Seal Inner Air Element, 152.4 mm I.D., 206.4 mm O.D., 1 ea
Catalog No. 5000508382
Encompass
This radial seal inner air element provides essential filtration for heavy-duty equipment applications. Designed to fit within specified outer air elements, it ensures reliable protection and optimal performance in demanding environments.
  • Compatible with Case-International and New Holland equipment
  • Features an inside diameter of 6 in. (152.4 mm)
  • Outside diameter measures 8 1/8 in. (206.4 mm)
  • Height of 12.96875 in. (329.4 mm)
  • Designed for use with outer air element RS5455
  • Replaces industry standard part numbers including Case 367350A1 and 87409407
Catalog No. 50-005-08382 Supplier MSC Supplier No. 55873434
This radial seal inner air element provides essential filtration for heavy-duty equipment applications. Designed to fit within specified outer air elements, it ensures reliable protection and optimal performance in demanding environments.
  • Compatible with Case-International and New Holland equipment
  • Features an inside diameter of 6 in. (152.4 mm)
  • Outside diameter measures 8 1/8 in. (206.4 mm)
  • Height of 12.96875 in. (329.4 mm)
  • Designed for use with outer air element RS5455
  • Replaces industry standard part numbers including Case 367350A1 and 87409407