Learn More
MSC Baldwin Filters Air Filter, Inner Element, Heavy Duty, 1 ea
Catalog No. 5000509839
Encompass
This inner heavy-duty air element is designed for engine air intake filtration to prevent contaminants from reaching the combustion chamber. It serves as a replacement component in compatible industrial air filtration systems, including those used in Komatsu excavators.
  • Fits inside PA3808-FN
  • Compatible with Komatsu 20501-K1480
  • Axial seal design
  • Includes one attached gasket and one additional G103-N gasket
  • Dimensions: 419.1 mm length, 115.9 mm outside diameter, and 16.7 mm and 87.3 mm inside diameters
  • Engineered for mobile engine applications
Catalog No. 50-005-09839 Supplier MSC Supplier No. 55865786
This inner heavy-duty air element is designed for engine air intake filtration to prevent contaminants from reaching the combustion chamber. It serves as a replacement component in compatible industrial air filtration systems, including those used in Komatsu excavators.
  • Fits inside PA3808-FN
  • Compatible with Komatsu 20501-K1480
  • Axial seal design
  • Includes one attached gasket and one additional G103-N gasket
  • Dimensions: 419.1 mm length, 115.9 mm outside diameter, and 16.7 mm and 87.3 mm inside diameters
  • Engineered for mobile engine applications