Learn More
MSC Baldwin Filters Engine Inner Air Filter, Round, 2.69 in. Diameter, 9.09 in. Length, 1 ea
Catalog No. 5000509803
Encompass
This heavy-duty inner air filter element is designed for use in engine air intake systems to effectively prevent contaminants from reaching the combustion chamber, supporting optimal engine performance and longevity in industrial and mobile applications.
  • Designed as a round inner air filter element
  • Includes a gasket for secure installation
  • Fits inside the PA3816 outer filter
  • Engineered for engine air intake filtration
  • Dimensions measure 23.1 cm (9 3/32 in.) in height and 6.8 cm (2 11/16 in.) in outside diameter
Catalog No. 50-005-09803 Supplier MSC Supplier No. 55865760
This heavy-duty inner air filter element is designed for use in engine air intake systems to effectively prevent contaminants from reaching the combustion chamber, supporting optimal engine performance and longevity in industrial and mobile applications.
  • Designed as a round inner air filter element
  • Includes a gasket for secure installation
  • Fits inside the PA3816 outer filter
  • Engineered for engine air intake filtration
  • Dimensions measure 23.1 cm (9 3/32 in.) in height and 6.8 cm (2 11/16 in.) in outside diameter