Learn More
MSC Baldwin Filters Radial Seal Air Filter Element, Inner, 108.0 mm OD, 169.1 mm Length, 1 ea
Catalog No. 5000509699
Encompass
The RS3537 is a radial seal inner heavy-duty air filter element designed to support rigorous engine applications. It ensures the correct functioning of equipment by preventing suspended contaminants from reaching the combustion chamber.
  • Radial seal design
  • Compatible with inner filter housings, including RS3536
  • Includes a 69.9 mm (2 3/4 in.) inner gasket on one end
  • Constructed for high-performance protection of engines and mobile equipment
  • Measures 108.0 mm (4 1/4 in.) in outer diameter and 169.1 mm (6 21/32 in.) in length
Catalog No. 50-005-09699 Supplier MSC Supplier No. 55874002
The RS3537 is a radial seal inner heavy-duty air filter element designed to support rigorous engine applications. It ensures the correct functioning of equipment by preventing suspended contaminants from reaching the combustion chamber.
  • Radial seal design
  • Compatible with inner filter housings, including RS3536
  • Includes a 69.9 mm (2 3/4 in.) inner gasket on one end
  • Constructed for high-performance protection of engines and mobile equipment
  • Measures 108.0 mm (4 1/4 in.) in outer diameter and 169.1 mm (6 21/32 in.) in length