Learn More
MSC Baldwin Filters, INNER HD AIR ELEMENT
Catalog No. 5000509654
Encompass
This inner air filter element is designed for heavy-duty filtration applications in engines and mobile equipment. It provides essential protection by capturing particulate matter, helping to maintain engine performance and longevity.
  • Fits equipment from manufacturers including Bomag and Deutz
  • Designed to serve as an inner element for multi-stage filtration assemblies
  • Constructed with an attached gasket for secure installation
  • Outside diameter of 84.1 mm (3 5/16 in.)
  • Height of 312.7 mm (12 5/16 in.)
  • Compatible with primary air filter element PA2423
Catalog No. 50-005-09654 Supplier MSC Supplier No. 55865877
This inner air filter element is designed for heavy-duty filtration applications in engines and mobile equipment. It provides essential protection by capturing particulate matter, helping to maintain engine performance and longevity.
  • Fits equipment from manufacturers including Bomag and Deutz
  • Designed to serve as an inner element for multi-stage filtration assemblies
  • Constructed with an attached gasket for secure installation
  • Outside diameter of 84.1 mm (3 5/16 in.)
  • Height of 312.7 mm (12 5/16 in.)
  • Compatible with primary air filter element PA2423