Learn More
Chemglass Life Sciences Inner Tube Only, Airfree, Schlenk
SDP
Catalog No. AF0513A22
Encompass_Preferred
Inner Tube Only, Airfree, Schlenk. Inner tube only. Replacement part for AF0513A21. Complete bubbler consists of the lower reservoir, adjustable inner tube, compression cap and Viton o-ring.
Catalog No. AF0513A22 Supplier Chemglass Life Sciences Supplier No. AF0513A22
Inner Tube Only, Airfree, Schlenk. Inner tube only. Replacement part for AF0513A21. Complete bubbler consists of the lower reservoir, adjustable inner tube, compression cap and Viton o-ring.