Learn More
Andwin Scientific INNOCULATING LOOP/8' HANDLE
SDP
Supplier:  Andwin Scientific 50815200
Encompass
The Inoculating Loops are attached to an insulated aluminum handle. The product has an overall length of 20.3 centimeters or 8 inches. It has an inner diameter of 2 millimeters. This product is sold individually.
Catalog No. NC9778211