Learn More
Chemglass Life Sciences Chemglass Inoculation Pick, Standard Needle Point, 0.254 cm OD x 6.35 cm, 1,600/Pack
SDP
Catalog No. NC2459109
Encompass_Preferred
Chemglass Life Sciences inoculation picks are needle-point wooden picks designed for precise colony transfer, streaking, and inoculation in microbiology workflows. Made from 100% USA-grown white birch in an ISO- and FDA-registered facility, these picks are supplied non-sterile and intended to be autoclaved prior to use.
  • Needle point tip for precise colony picking.
  • 0.254 cm outer diameter and 6.35 cm length.
  • Pack of 1,600 picks for high-volume workflows.
  • Made from 100% USA-grown white birch.
  • Non-sterile; autoclave before use.
  • Made in the USA in an ISO- and FDA-registered facility.
Catalog No. NC2459109 Supplier Chemglass Life Sciences Supplier No. CLS3670106
Chemglass Life Sciences inoculation picks are needle-point wooden picks designed for precise colony transfer, streaking, and inoculation in microbiology workflows. Made from 100% USA-grown white birch in an ISO- and FDA-registered facility, these picks are supplied non-sterile and intended to be autoclaved prior to use.
  • Needle point tip for precise colony picking.
  • 0.254 cm outer diameter and 6.35 cm length.
  • Pack of 1,600 picks for high-volume workflows.
  • Made from 100% USA-grown white birch.
  • Non-sterile; autoclave before use.
  • Made in the USA in an ISO- and FDA-registered facility.