Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass Inoculation Pick, Standard, Semi Point - 0.254 cm OD x 6.35 cm, 1,600/Pack
SDP

Catalog No. NC2459107
Encompass_Preferred

Single-use wooden inoculation picks designed for routine microbiology workflows. The semi-point profile provides reliable sample transfer while minimizing disruption to agar surfaces. Supplied in bulk packs for high-volume laboratories.

  • Material: 100% USA-grown white birch.
  • Dimensions: 0.254 cm outer diameter x 6.35 cm length.
  • Pack quantity: 1,600 per pack.
  • Sterility: non-sterile.
  • Compliance: meets FDA requirements for food contact.
  • Environmental: naturally biodegradable.
  • Manufacturing: made in the USA in an ISO- and FDA-registered facility.

Catalog No. NC2459107 Supplier Chemglass Life Sciences Supplier No. CLS3670104
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Single-use wooden inoculation picks designed for routine microbiology workflows. The semi-point profile provides reliable sample transfer while minimizing disruption to agar surfaces. Supplied in bulk packs for high-volume laboratories.

  • Material: 100% USA-grown white birch.
  • Dimensions: 0.254 cm outer diameter x 6.35 cm length.
  • Pack quantity: 1,600 per pack.
  • Sterility: non-sterile.
  • Compliance: meets FDA requirements for food contact.
  • Environmental: naturally biodegradable.
  • Manufacturing: made in the USA in an ISO- and FDA-registered facility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.