Learn More
Fujirebio Diagnostics Inc INNOTEST HTAU ELISA Kit, Total Tau protein, 96 Tests, Color-coded components, 25 μL CSF minimum sample volume, Calibrator range: 50-2500 pg/mL
Supplier:  Fujirebio Diagnostics Inc 81579
Encompass
INNOTEST HTAU ELISA Kit, Total Tau protein, 96 Tests, Color-coded components, 25 μL CSF minimum sample volume, Calibrator range: 50-2500 pg/mL
Catalog No. NC0869787