Learn More
Innovative Research Inc INNOV GRD US ORIGN BOV SYN FL
Catalog No. NC3037468
Encompass_Preferred
Innovative Grade US Origin Bovine Synovial Fluid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States ensuring that only the highest-quality material is provided.Synovial Fluid is collected from the joint using a syringe and is filtered through gauze.
Catalog No. NC3037468 Supplier Innovative Research Inc Supplier No. IGBOSYNZ100ML
Innovative Grade US Origin Bovine Synovial Fluid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States ensuring that only the highest-quality material is provided.Synovial Fluid is collected from the joint using a syringe and is filtered through gauze.