Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Guard Cartridge 10 x 10mm

Supplier:  Phenomenex Inc IX9501102S

Encompass

Innoval 5m ODS-2 100 IX950110-2S

Catalog No. 50-239-7679


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.