Learn More
Siemens Healthineers INNOVANCE™ D-Dimer Controls
Catalog No. 23044673 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
L1 (5 x 1 mL), L2 (5 x 1 mL)
Catalog No. Quantity
23-044-673 L1 (5 x 1 mL), L2 (5 x 1 mL)
1 options
This item is not returnable. View return policy
Catalog No. 23-044-673 Supplier Siemens Healthineers Supplier No. 10446006
Add to Cart
Add to Cart
This item is not returnable. View return policy
INNOVANCE™ D-Dimer Control 1 and Control 2 are assayed, normal and pathological level, intralaboratory quality controls for assessment of precision and analytical bias in the quantitative determination of D-dimer on Siemens and Sysmex™ Systems.
INNOVANCE™ D-Dimer Control 1 and 2 are assayed controls for the assessment of precision and analytical bias in the normal and pathological range for the determination of D-Dimer on Sysmex™ systems.
TRUSTED_SUSTAINABILITY
Specifications
Type D-Dimer Control Kit
For Use With (Application) D-Dimer Testing
Level 1 to 2
Quantity L1 (5 x 1 mL), L2 (5 x 1 mL)