Learn More
Siemens Healthineers INNOVANCE™ D-Dimer Diluent
Catalog No. 23044734 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 5 mL
Catalog No. Quantity
23-044-734 10 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-734 Supplier Siemens Healthineers Supplier No. 10487039
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in conjunction with INNOVANCE D-Dimer to dilute samples.
The INNOVANCE™ D-Dimer Diluent is used for dilution of elevated D-Dimer concentrations with the INNOVANCE™ D-Dimer assay.
TRUSTED_SUSTAINABILITY
Specifications
Type Diluent
For Use With (Application) D-Dimer Testing
Quantity 10 x 5 mL