Learn More
Siemens Healthineers INNOVANCE™ D-Dimer Kit
Catalog No. 23044669 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
150 Tests/Kit
300 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-669 150 Tests/Kit
23-044-670 300 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-669 Supplier Siemens Healthineers Supplier No. 10445981
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the quantitative determination of cross-linked fibrin degradation products (D-dimers) in human plasma on Siemens Sysmex™ Coagulation Systems.
Combining an excellent clinical sensitivity of >98.9% with a high negative predictive value (NPV) of 99%, the INNOVANCE™ D-Dimer assay's FDA-cleared exclusion claim gives clinicians greater confidence that translates into better patient management.
The fully automated INNOVANCE™ D-Dimer assay offers consistent performance on Siemens Healthineers and Sysmex coagulation instrument platforms for equivalent D-dimer test results. Speed and performance make INNOVANCE™ D-Dimer a robust, cost-effective assay for both routine and emergency use.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) D-Dimer Testing
Quantity 150 Tests/Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov