Learn More
Siemens Healthineers INNOVANCE™ Heparin Calibrators (US)
Catalog No. 23044769 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
5 x 1 mL (1 for each level)
Catalog No. Quantity
23-044-769 5 x 1 mL (1 for each level)
1 options
This item is not returnable. View return policy
Catalog No. 23-044-769 Supplier Siemens Healthineers Supplier No. 10873530
Add to Cart
Add to Cart
This item is not returnable. View return policy
For calibration of the INNOVANCE™ Heparin assay.
The INNOVANCE™ Heparin Calibrator consists of 5 levels.The calibrator levels are used to establish a reference curve which then can be used to quantify the heparin activity of UFH and LMWH containing plasmas.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Heparin Testing
Level 1 to 5
Quantity 5 x 1 mL (1 for each level)