Learn More
Siemens Healthineers INNOVANCE™ Heparin LMW Controls (US)
Catalog No. 23044773 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
Control 1 - Low
Control 2 - High
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-044-773 Control 1 - Low
23-044-772 Control 2 - High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-773 Supplier Siemens Healthineers Supplier No. 10873534
Add to Cart
Add to Cart
This item is not returnable. View return policy
For quality control of the INNOVANCE™ Heparin assay.
INNOVANCE™ Heparin LMW Controls are used to monitor the performance of low molecular weight heparin with the INNOVANCE™ Heparin assay.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Heparin Testing
Level Control 1 - Low
Quantity 5 x 1 mL