Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Acid Rain and Environment: Acidity and Plant Grow Kit
GREENER_CHOICESCI_ED

Catalog No. S05883 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05883 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05883 Supplier Innovating Science™ Supplier No. IS9704
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this activity, students will examine detrimental effects of acidic conditions on plants.

  • Plants will be grown under normal soil conditions, mildly acidic soil conditions, and very acidic soil conditions
  • Students will determine if acidity has any impact on the growth of the plant
  • Kit contains enough materials for 15 groups

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To determine acidity on growth of plant
Includes 4 x 30ml hydrochloric acid 12m un1789, potting soil, 45 5 oz. translucent cup, 45 5 oz. translucent cup, 45 pieces absorbent wick pad, 1 radish seeds
Quantity 1
Class Size 15 Lab Groups
Product Type Acidity and Plant Growth Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.