Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Agarose Gel Electrophoresis Reagent Pack
SCI_ED

Catalog No. S20655 Shop All Aldon Products
Change view
Click to view available options
Quantity:
11
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20655 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20655 Supplier Innovating Science™ Supplier No. IS5250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 500 mL TBE buffer (5X)
  • 5 g Agarose, powdered
  • 60 mL DNA Stain (20X)
  • 10 mL Loading Dye (10X)

All the necessary reagents to prepare and run agarose gels in one convenient package.

Contains: TBE Buffer 5X, 500 mL Agarose Powder, 5 g DNA Stain 20X, 60 mL Loading Dye 10X, 10 mL

Specifications

Detection Method Fluorescence
Concentration 5x
Format Bottle
Sample Type Cell Extracts, Immunoprecipitates, Purified Enzymes
For Use With (Application) Electrophoresis
For Use With (Equipment) Electrophoresis
Includes 500 mL TBE buffer (5X), 5 g Agarose, powdered, 60 mL DNA Stain (20X), 10 mL Loading Dye (10X)
Label or Dye Dye
Product Type Kit
Quantity 11
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.