Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 13: Enzyme Activity
GREENER_CHOICESCI_ED

Catalog No. s07044 Shop All Aldon Products
Change view
Click to view available options
:
Enzyme activity kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07044 Enzyme activity kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07044 Supplier Innovating Science™ Supplier No. IS3713
Only null left
Add to Cart
Add to Cart

Includes

  • Guaiacol concentrate - 1.5mL
  • pH 3 Buffer - 100mL
  • pH 5 Buffer - 100mL
  • pH 6 Buffer - 100mL
  • pH 7 Buffer - 100mL
  • pH 8 Buffer - 100mL
  • pH 10 Buffer - 100mL
  • Hydrogen peroxide - 1.0%
  • Teacher's manual and student study guide

Requires

  • Distilled or deionized water
  • Parafilm or similar
  • Test tube - 16 x 150mm
  • 16 graduated pipettes - 1mL
  • 16 graduated pipettes - 10mL
  • Eight graduated pipettes - 5mL
  • Eight pipettes or pipette bulbs
  • Eight test tube racks
  • Eight timers or stopwatches
Recommended: Spectrophotometer

Students will use turnip extracts to learn the nature and specificity of enzyme-catalyzed reactions.

The reaction between hydrogen peroxide and perioxidase enzyme will be examined after testing with one or more influential factors.

  • Kits contains materials for eight groups
  • Meets AP Science Practices 4, 5, 6 and 7, and Big Idea 4
  • For Grades 10-12

Specifications

Certifications/Compliance Meets AP science practices 4, 5, 6 and 7 and big idea 4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Students will use turnip extracts to learn the nature and specificity of enzyme-catalyzed reactions, materials for eight lab groups
Includes 1.5ml concentrate guaiacol, ph 3 buffer, ph 5 buffer, ph 6 buffer, ph 7 buffer, ph 8 buffer and ph 10 buffer of 100ml, 50ml 1.0% hydrogen peroxide, teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 8 Lab Groups
Product Type AP Biology Investigation Kit #13: Enzyme Activity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.