Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 6: Cellular Respiration
GREENER_CHOICESCI_ED

Catalog No. s07057 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07057 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause and reproductive harm.
Catalog No. S07057 Supplier Innovating Science™ Supplier No. IS3706
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause and reproductive harm.

Requires

  • Eight graduated cylinders (100mL)
  • Eight thermometers
  • Eight stopwatches or digital timers
  • Ice
  • Wax marking pencils or markers
  • Paper towels
  • Masking tape

Students measure and record the rate of oxygen consumption (cellular respiration).

  • Activity involves use of a respiration chamber
  • Germinating seeds and non-germinating material are used as control samples
  • Meets AP Science Practices 1, 2, 3, 6 and 7 and Big Idea 2
  • This package conforms to 49 CFR 173.4 for domestic highway or rail transport only
  • For eight lab groups
  • Kit Includes:
    • Pea Seeds (1 Pack)
    • Plastic Beads (1 Pack)
    • 15% Potassium Hydroxide (1 Bottle)
    • Non-absorbent, absorbent cotton (1 Pack)
    • Petroleum jelly sealant (1 Tube)
    • 8 trays
    • 48 washers
    • 24 rubber stoppers
    • 24 Respiration Chambers
    • 24 Graduated Pipets
    • Teacher's manual
    • Student study guides

Specifications

Certifications/Compliance 49 CFR 173.4, meets AP Science Practices 1, 2, 3, 6 and 7 along with Big Idea 2
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Measuring and recording the rate of oxygen consumption with germinating versus non-germinating samples using respiration chamber
Quantity 1 Kit
Dimensions (L x W x H) 29.25 x 16.5 x 5 in.
Class Size 8 Lab Groups
Product Type AP Biology Investigation Kit #6: Cellular Respiration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.