Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 7: Cell Division: Mitosis and Meiosis
GREENER_CHOICESCI_ED

Catalog No. s07062 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07062 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which are known to the State of California to cause cancer.
Catalog No. S07062 Supplier Innovating Science™ Supplier No. IS3707
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which are known to the State of California to cause cancer.

Students will study the cellular steps involved in DNA replication and cell division in both mitosis and meiosis.

  • Stages of mitosis in the preparation of plant root tips
  • Crossing over in meiosis via Sordaria cultures
  • Meets AP Science Practices 1, 5, 6, and 7, and Big Idea 3
  • Teacher's manual and student's guide included
  • Materials for 32 students (8 groups of 4)
  • Kit Includes:
    • 6.0M Hydrochloric Acid (30 mL)
    • 1% Toluidine blue (30 mL)
    • Sand (500 g)
    • Sordaria Agar (200 mL)
    • Sordaria Mating Agar (200 mL)
    • Slides, Coverslips (1 Pack Ea.)
    • 8 forceps
    • 16 Plastic pipettes
    • 20 Petri dishes (sterile)
    • 8 Inoculating loops (sterile, disposable)
    • 7 oz. cups (8)
    • 10 Disposable scalpels
    • Coupon for perishable materials

Specifications

Certifications/Compliance 49 CFR 173.4, meets AP science practices 1, 5, 6, and 7 along with Big Idea 3
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Studying the cellular steps involved in DNA replication and cell division in both mitosis and meiosis
Quantity 1 Kit
Dimensions (L x W x H) 12.625 x 9.5 x 6.75 in.
Class Size 32 Students (8 Groups)
Product Type AP Biology Investigation Kit #7: Cell Division: Mitosis and Meiosis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.