Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 8: Bacterial Transformation
GREENER_CHOICESCI_ED

Catalog No. s07058 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07058 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07058 Supplier Innovating Science™ Supplier No. IS3708
Only null left
Add to Cart
Add to Cart

Includes

  • Sterile Calcium Chloride 0.05M (30mL)
  • 16 Sterile volumetric pipets
  • Eight Pipet bulbs
  • Eight micropipets w/plunger (10μL)
  • 16 Microcentrifuge Tubes (1.5mL)
  • Eight sterile inoculating loops
  • 40 Petri dishes
  • Four Luria agar (200mL)
  • One Luria broth tube (9mL)
Perishable materials are supplied via coupon redeemable by fax, phone or e-mail, and include:
  • One E. Coli culture (freeze dried)
  • Two bottles 0.02g Ampicillin
  • 1000ng pUC 19 Plasmid
  • One Tryptic Soy Broth tube (9mL)
  • One Tryptic Soy Agar slant (6mL)
  • One sterile, disposable pipet

Students genetically engineer bacteria through transformation with a plasmid that confers antibiotic resistance.

 

  • Efficiency of the procedure is determined by examining the number of resistant bacteria
  • Includes teachers manual and student study guide copymasters
  • Meets AP Science Practices 1, 3, 5, 6, and 7, and Big Idea 3
  • For eight lab groups

 

Recommended: Incubator (37°C). Requires: Large beaker, microwave (not necessary if melting agar in a waterbath), microfuge tube racks, hot plate or waterbath, markers, and container of ice.

Specifications

Certifications/Compliance Meets AP science practices 1, 3, 5, 6, and 7 and Big Idea 3
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Students genetically engineer bacteria through transformation with a plasmid that confers antibiotic resistance
Includes 30ml 0.05m sterile calcium chloride, 16 sterile volumetric pipets, 8 pipet bulbs, 8 x 10μl micropipets with plunger, 16 x 1.5ml microcentrifuge tubes, 8 sterile inoculating loops, 40 petri dishes, 4 x 200ml luria agar, 9ml luria broth tube and coupon for perishable supplies
Quantity 1
Dimensions (L x W x H) 12.125 x 9.25 x 6.5 in.
Class Size 8 Lab Groups
Product Type AP™ Biology Investigation Kit #8: Biotechnology: Bacterial Transformation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.