Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 9: Restriction Enzyme Analysis
GREENER_CHOICESCI_ED

Catalog No. S07059 Shop All Aldon Products
Change view
Click to view available options
Class Size:
8 Lab Groups or 32 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07059 8 Lab Groups or 32 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07059 Supplier Innovating Science™ Supplier No. IS3709
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Prepared agarose, 0.8%, 200mL TBE buffer, 5X, 500mL
  • DNA Stain, 20X, 60mL DNA Samples, 100μL each:
  • DNA Marker (Lambda DNA HindIII digest)
  • Sample #1 (Lambda DNA)
  • Sample #2 (Lambda DNA BstEII digest)
  • Sample #3 (Lambda DNA EcoRI digest)
  • Sample #4 (Lambda DNA BstEII digest)

Requires

  • Agarose electrophoresis chamber
  • Microwave or hot water bath
  • Electrophoresis power supply
  • Distilled or deionized water
  • Micropipettes w/micropipette tips, 10μL
  • Gel staining trays or similar (large enough container to accommodate flat agarose gels)

In this lab students will understand the action and specificity of restriction enzymes, while learning the components involved in the process of DNA electrophoresis and the role of each component as it applies to the entire process of electrophoresis.

Students will perform the electrophoresis process on DNA samples treated with different restriction enzymes and construct a standard curve using a known DNA sample while determining the approximate size of the DNA fragments in unknown samples. Meets AP Science Practices 3 and 6, and Big Idea 3. Materials provided for eight lab groups.
 

Specifications

Certifications/Compliance Meets AP science practices 3 and 6 and big idea 3
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Students will understand the action and specificity of restriction enzymes
Includes Prepared agarose, 0.8%, 200mL;  TBE buffer, 5X, 500mL; DNA Stain, 20X, 60mL; DNA Samples, 100μL each: DNA Marker (Lambda DNA HindIII digest), Sample 1 (Lambda DNA), Sample 2 (Lambda DNA BstEII digest), Sample 3 (Lambda DNA EcoRI digest), Sample 4 (Lambda DNA BstEII digest)
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 8 Lab Groups or 32 Students
Product Type AP Biology Investigation Kit #9: Biotechnology: Restriction Enzyme Analysis of DNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.