Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Analysis of Potassium Aluminum Sulfate 12 Hydrate
SCI_ED

Catalog No. S97016 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S97016 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97016 Supplier Innovating Science™ Supplier No. IS8029
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 100g Aluminum Potassium Sulfate (2), 25g Barium Nitrate UN1446 (4), teachers guide and student copy masters

Use analytical tests to confirm the correct compound has been produced following synthesis.

Confirms that the crystals formed are in fact Potassium Aluminum Sulfate 12 hydrate by comparing the melting point to published results and determining the percent of sulfate in the compound.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Analysis of potassium aluminum sulfate 12 hydrate formed by comparing the melting point to published results
Includes 2 x 100g aluminum potassium sulfate, 4 x 25g barium nitrate
Quantity 1
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Type Analysis of Potassium Aluminum Sulfate 12 Hydrate Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.