Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Beer-Lambert Law
GREENER_CHOICESCI_ED

Catalog No. S90735 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90735 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90735 Supplier Innovating Science™ Supplier No. IS8010
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 50mL Ferric Chloride solution (4); 25g L+Ascorbic Acid (Vitamin C) (2); 100mL 1,10-Phenanthroline solution (2); 200mL Acetate Buffer solution (2); teacher's guide; and student copy masters

Use of the spectrophotometric assay for iron to prepare and analyze a series of five solutions with different concentrations of iron.

The probability that a photon of light will be adsorbed by a solution is directly proportional to the concentration of the absorbing molecules, the thickness of the sample and the degree to which the molecules absorb light. This relationship is called the Beer-Lambert Law and is used in analytical chemistry to determine concentrations of various compounds spectrophotometrically.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Used in analytical chemistry to determine concentrations of various compounds spectrophotometrically
Includes 1 x 25ml 0.1m ferric chloride, 1 x 5g ascorbic acid, 1 x 50ml 1,10 phenanthroline, 1 x 100ml acetate buffer, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.1875 x 2.625 x 8.25 in.
Class Size 15 Lab Groups
Product Type Beer-Lambert Law Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.