Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Complete Set of 22
GREENER_CHOICESCI_ED

Catalog No. s90748 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90748 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Potassium Bromate, Methanol/ Methyl Isobutyl Ketone, Phenolphthalein, Strong Inorganic Acid Mists containing Sulfuric Acid, Naphthalene, p-Dichlorobenzene, Toluene, and Mercury/ Mercury Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S90748 Supplier Innovating Science™ Supplier No. IS8023
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, Potassium Bromate, Methanol/ Methyl Isobutyl Ketone, Phenolphthalein, Strong Inorganic Acid Mists containing Sulfuric Acid, Naphthalene, p-Dichlorobenzene, Toluene, and Mercury/ Mercury Compounds, which are known to the State of California to cause cancer and reproductive harm.

Be prepared for the entire school year with this complete set of all 22 AP Chemistry lab activities.

Complete set includes all 22 AP Chemistry Kits:
  • Thermochemistry & Hess Law
  • Activity Series
  • Mole Ratio Reactants
  • Le Chatelier's Principle
  • Ester Formation
  • Acid-Base
  • Reaction Kinetics
  • Dissociation of Weak Acids
  • Qualitative Analysis
  • Beer-Lambert Law
  • Stoichiometry
  • Equilibrium Constant
  • Oxidation-Reduction
  • Freezing Point Depression
  • Vapor Pressure
  • Electrochemical Cells
  • Thin-Layer Chromatography
  • Molecular Mass of a Liquid
  • Thermochromism
  • Water Hardness
  • Synthesis of Aspirin
  • Grignard Method

Specifications

Grade 9 to 12
For Use With (Application) For lab activities
Includes Thermochemistry & Hess Law, Activity Series, Mole Ratio Reactants, Le Chatelier's Principle, Ester Formation, Acid-Base, Reaction Kinetics, Dissociation of Weak Acids, Qualitative Analysis, Beer-Lambert Law, Stoichiometry, Equilibrium Constant, Oxidation-Reduction, Freezing Point Depression, Vapor Pressure, Electrochemical Cells, Thin-Layer Chromatography, Molecular Mass of a Liquid, Thermochromism, Water Hardness, Synthesis of Aspirin, Grignard Method
Class Size 15 Lab Groups
Product Type Complete Set Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.