Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Determination of the Empirical Formula of Magnesium Oxide
SCI_ED

Catalog No. S97012 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S97012 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97012 Supplier Innovating Science™ Supplier No. IS8025
Add to Cart
Add to Cart

Includes

Materials for 15 groups: One roll of magnesium metal ribbon UN1869, teachers guide and student copy masters

Illustrates the law of conservation of mass and the law of constant composition.

  • To study the quantitative stoichiometric relationships governing mass and amount using the combustion reaction of magnesium metal
  • Magnesium reacts with oxygen from the air in a crucible
  • Measures mass before and after oxidation
  • Use results to calculate the experimental empirical formula and compare to the theoretical empirical formula

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To study the quantitative stoichiometric relationships governing mass and amount using the combustion reaction of magnesium metal
Includes 1 roll magnesium metal, ribbon, Teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Type Determination of the Empirical Formula of Magnesium Oxide Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.