Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Mole Ratio
SCI_ED

Catalog No. S90728 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90728 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90728 Supplier Innovating Science™ Supplier No. IS8003
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 400g Sodium Thiosulfate, Anhydrous; 250mL Sodium Hypochlorite, 13% conc. UN1791; teacher's guide; and student copy masters

In this experiment, the total number of moles of reactants are kept constant while varying each reactant.

The measurements are made on each different ratio until the optimum ratio — the stoichiometric ratio in the equation — is made, which consumes the greatest amount of reactants, produces the greatest amount of product and produces the greatest amount of heat.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes 3 x 250ml sodium hypochlorite 13% concentrate, 1 x 237g sodium thiosulfate, anhydrous, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 7.25 x 7.25 x 8.375 in.
Class Size 15 Lab Groups
Product Type Mole Ratio Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.