Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Oxidation-Reduction
GREENER_CHOICESCI_ED

Catalog No. S90738 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S90738 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
Catalog No. S90738 Supplier Innovating Science™ Supplier No. IS8013
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Includes

Materials for 15 groups: 10g Ferrous Ammonium Sulfate; 100mL Sulfuric Acid, 6M solution UN2796; 100mL Potassium Thiocyanate, 1.0M solution (2); 25mL Potassium Permanganate, 2% solution (2); 100mL Tin(II) Chloride, 0.1M solution (3); 25mL Hydrogen Peroxide, 3% (2); 25mL Methylene Blue, 1% solution; 250mL Potassium Hydroxide, 1.0M solution UN1814 (3); 50g Dextrose Monohydrate (2); 100g glucose; teacher's guide; and student copy masters

In this series of experiments, the oxidation-reduction reactions of compounds are monitored via a change in their optical characteristics.

A compound that is colorless in solution when reduced is converted to a deeply colored solution when oxidized. The complete balanced reactions for each step should be written showing the transfer of electrons during oxidation and reduction.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To study the oxidation-reduction reactions of compounds are monitored
Includes 1 x 10g ferrous ammonium sulfate, 1 x 100ml 6m sulfuric acid, 2 x 100ml potassium thiocyanate 1m, 1 x 25ml potassium permanganate, 1 x 25ml hydrogen peroxide, 3%, 3 x 100ml 0.1m stannous chloride, 1 x 25ml 1% methylene blue, 3 x 250ml potassium hydroxide
Quantity 1
Dimensions (L x W x H) 8 x 8 x 8 in.
Class Size 15 Lab Groups
Product Type Oxidation-Reduction Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.