Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Reaction Kinetics
GREENER_CHOICESCI_ED

Catalog No. s90732 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S90732 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90732 Supplier Innovating Science™ Supplier No. IS8007
Only null left
Add to Cart
Add to Cart

Includes

Materials for 15 groups: 100mL Potassium Iodide, 0.1% solution 0.006M (2); 100mL Sodium Thiosulfate, 0.001M solution (2); 100mL Hydrogen Peroxide, 3% solution 0.88M (2); 5 capsules Starch Soluble (5 caps per bottle); 100mL Hydrochloric Acid, 0.1M solution UN1789 (2); 150 pipets; teacher's guide; and student copy masters

In this experiment, the time it takes for a reaction to occur is monitored by observing a color change.

By varying concentration of reactants, the rate law is determined. By varying the temperature, the activation energy is determined. The data should be graphed and analyzed during the lab so additional measurements can be made if necessary.
In a chemical reaction, reactants are converted to product at a given rate. This rate can be changed by alternating either the temperature of the reaction, the form of the reactants, the concentration of reactants or products, or by adding a catalyst.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) By varying concentration of reactants, the rate law is determined, by varying the temperature, the activation energy is determined
Includes 2 x 100mL Potassium Iodide, 0.1% solution (0.006M), 2 x 100mL Sodium Thiosulfate, 0.001M solution, 2 x 100mL Hydrogen Peroxide, 3% solution (0.88M), 5 capsules Starch soluble, 2 x 100mL Hydrochloric Acid, 0.1M solution UN1789 pipettes, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 8 x 8 x 8 in.
Class Size 15 Lab Groups
Product Type Reaction Kinetics Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.